α,α,α-Trifluoro-o-cresol
Supplier: Thermo Scientific
Synonyms: 2-Hydroxybenzotrifluoride, 2-(Trifluoromethyl)phenol
SB01911.1 SB01911.10 SB01911.50
MAYBSB01911.1EA 0 EUR
MAYBSB01911.1 MAYBSB01911.10 MAYBSB01911.50
α,α,α-Trifluoro-o-cresol
α,α,α-Trifluoro-o-cresol
Formula: C₇H₅F₃O
Boiling Pt: 147 °C (1013 hPa)
Melting Pt: 46 °C
Density: >1 g/cm³ (20 °C)
MDL Number: MFCD00002222
CAS Number: 444-30-4
EINECS: 207-148-7
UN: 3261
ADR: 8,III