α,α,α-Trifluoro-m-cresol
Supplier: Thermo Scientific
Warning
Synonyms: 3-(Trifluoromethyl)phenol, 3-Hydroxybenzotrifluoride
SB01203.1 SB01203.50
MAYBSB01203.1EA 0 EUR
MAYBSB01203.1 MAYBSB01203.50
α,α,α-Trifluoro-m-cresol
α,α,α-Trifluoro-m-cresol
Formula: C₇H₅F₃O
MW: 162.11 g/mol
Boiling Pt: 178
Melting Pt: –2 °C
Density: 1.34 g/cm³ (20 °C)
Flash Pt: 73
Storage Temperature: Ambient
MDL Number: MFCD00002299
CAS Number: 98-17-9
EINECS: 202-645-5
UN: 3265
ADR: 8 III