α,α,α-Trifluoro-m-cresol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 3-(Trifluoromethyl)phenol, 3-Hydroxybenzotrifluoride
MQ100
156035-100G 156035-25G
SIAL156035-100GEA 89.1 EUR
SIAL156035-100G SIAL156035-25G
α,α,α-Trifluoro-m-cresol, Sigma-Aldrich®
α,α,α-Trifluoro-m-cresol
Formula: C₇H₅F₃O
MW: 162.11 g/mol
Boiling Pt: 178
Melting Pt: –2 °C
Density: 1.34 g/cm³ (20 °C)
Flash Pt: 73
Storage Temperature: Ambient
MDL Number: MFCD00002299
CAS Number: 98-17-9
EINECS: 202-645-5
UN: 3265
ADR: 8 III