1-tert-Butyl-4-iodobenzene, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-tert-Butyliodbenzol, 1-(tert-Butyl)-4-iodobenzene, 4-tert-Butyliodobenzene

M-Clarity™ quality level = MQ100

411205-1G 411205-5G
SIAL411205-1GEA 29.3 EUR
SIAL411205-1G SIAL411205-5G
1-tert-Butyl-4-iodobenzene, Sigma-Aldrich®
1-tert-Butyl-4-iodobenzene
Formula: C₁₀H₁₃I
MW: 260.12 g/mol
Boiling Pt: 114 °C (0.05 mmHg)
Density: 1.4392 g/cm³ (14 °C)
MDL Number: MFCD00052339
CAS Number: 35779-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR