4-Iodotoluene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Iodo-4-methylbenzene
M-Clarity™ quality level = MQ100
206555-100G 206555-25G
SIAL206555-100GEA 69.9 EUR
SIAL206555-100G SIAL206555-25G
4-Iodotoluene, Sigma-Aldrich®
4-Iodotoluene
Formula: C₇H₇I
Boiling Pt: 211.5 °C (1013 hPa)
Melting Pt: 33…35 °C
Density: 1.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001059
CAS Number: 624-31-7
EINECS: 210-841-7