(4-Iodophenyl)acetonitrile, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-(4-Iodophenyl)acetonitrile, 4-Iodobenzyl cyanide, 4-Iodophenylacetonitrile

M-Clarity™ quality level = MQ100

587028-1G 587028-5G
SIAL587028-1GEA 39.1 EUR
SIAL587028-1G SIAL587028-5G
(4-Iodophenyl)acetonitrile, Sigma-Aldrich®
(4-Iodophenyl)acetonitrile
Formula: C₈H₆IN
MW: 243.05 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 56…57 °C
MDL Number: MFCD00060306
CAS Number: 51628-12-7
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR