4'-Iodoacetophenone, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-(4-Iodophenyl)ethan-1-one, p-Iodoacetophenone, 1-(4-Iodophenyl)ethanone
M-Clarity™ quality level = MQ100
357804-25G 357804-5G
SIAL357804-25GEA 120 EUR
SIAL357804-25G SIAL357804-5G
4'-Iodoacetophenone, Sigma-Aldrich®
4'-Iodoacetophenone
Formula: C₈H₇IO
MW: 246.05 g/mol
Boiling Pt: 145…150 °C (20 mmHg)
Melting Pt: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD00045320
CAS Number: 13329-40-3
EINECS: 236-372-8