3-Iodo-p-toluic acid, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
278858-25G 278858-5G
SIAL278858-25GEA 148 EUR
SIAL278858-25G SIAL278858-5G
3-Iodo-p-toluic acid, Sigma-Aldrich®
3-Iodo-p-toluic acid
Formula: C₈H₇IO₂
MW: 262.05 g/mol
Boiling Pt: 329 °C (1013 hPa)
Melting Pt: 210…211 °C
MDL Number: MFCD00010392
CAS Number: 82998-57-0