2-Iodobenzyl alcohol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
183636-10G
183636-50G
SIAL183636-10GEA
79.3
EUR
SIAL183636-10G
SIAL183636-50G
2-Iodobenzyl alcohol, Sigma-Aldrich®
2-Iodobenzyl alcohol
Formula:
C₇H₇IO MW: 234.04 g/mol Boiling Pt: 145 °C (10 mmHg) Melting Pt: 91 °C |
MDL Number:
MFCD00004610 CAS Number: 5159-41-1 EINECS: 225-933-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...