Benzyl isothiocyanate
Supplier: Thermo Scientific
Synonyms: Benzylisothiocyanate
TL01140.1 TL01140.10 TL01140.50
MAYBTL01140.1EA 0 EUR
MAYBTL01140.1 MAYBTL01140.10 MAYBTL01140.50
Benzyl isothiocyanate
Benzyl isothiocyanate
Formula: C₈H₇NS
MW: 149.22 g/mol
Boiling Pt: 243
Melting Pt: 24
Density: 1.084 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004819
CAS Number: 622-78-6
EINECS: 210-753-9
UN: 2206