Benzylamine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: α-Aminotoluene
M-Clarity™ quality level = MQ200
407712-100ML 407712-800ML
SIAL407712-100MLEA 90.8 EUR
SIAL407712-100ML SIAL407712-800ML
Benzylamine, Sigma-Aldrich®
Benzylamine
Formula: (C₆H₅)CH₂NH₂
MW: 107.16 g/mol
Boiling Pt: 185 °C (1013 hPa)
Melting Pt: 10 °C
Density: 0.98 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00008106
CAS Number: 100-46-9
EINECS: 202-854-1
UN: 2735
ADR: 8,II
Merck Index: 13,01127