2-Ethylaniline, Sigma-Aldrich®
Supplier: Merck
Warning
MQ200
E11803-100G E11803-250G
SIALE11803-100GEA 35.9 EUR
SIALE11803-100G SIALE11803-250G
2-Ethylaniline, Sigma-Aldrich®
2-Ethylaniline
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 210
Melting Pt: –46 °C
Density: 0.983 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007751
CAS Number: 578-54-1
EINECS: 209-424-2
UN: 2273
ADR: 6.1 III