o-Toluidine, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 2-Toluidine, 2-Aminotoluene, 2-Methylaniline
M-Clarity™ quality level = MQ100
185426-100G 185426-500G 185426-5G
SIAL185426-100GEA 57.8 EUR
SIAL185426-100G SIAL185426-500G SIAL185426-5G
o-Toluidine, Sigma-Aldrich®
o-Toluidine
Formula: C₇H₉N
MW: 107.16 g/mol
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: < –15 °C
Density: 0.998 g/cm³ (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00007730
CAS Number: 95-53-4
EINECS: 202-429-0
UN: 1708
ADR: 6.1,II
Merck Index: 12,09674