4-tert-Butylaniline, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-(tert-Butyl)aniline
M-Clarity™ quality level = MQ100
209864-25G 209864-5G
SIAL209864-25GEA 209 EUR
SIAL209864-25G SIAL209864-5G
4-tert-Butylaniline, Sigma-Aldrich®
4-tert-Butylaniline
Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: 15 °C
Density: 0.941 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007899
CAS Number: 769-92-6
EINECS: 212-215-9
UN: 2735
ADR: 8,III