4-Butylaniline, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Butylbenzenamine
M-Clarity™ quality level = MQ100
112666-100G 112666-500G
SIAL112666-100GEA 81.6 EUR
SIAL112666-100G SIAL112666-500G
4-Butylaniline, Sigma-Aldrich®
4-Butylaniline
Formula: C₁₀H₁₅N
MW: 149.24 g/mol
Boiling Pt: 242 °C (1013 hPa)
Melting Pt: –14 °C
Density: 0.945 g/cm³ (20 °C)
MDL Number: MFCD00007925
CAS Number: 104-13-2
EINECS: 203-177-4
UN: 2810
ADR: 6.1,III