4-Ethylaniline, Sigma-Aldrich®
Supplier: Merck
MQ100
E12001-100ML E12001-500ML
SIALE12001-100MLEA 77.7 EUR
SIALE12001-100ML SIALE12001-500ML
4-Ethylaniline, Sigma-Aldrich®
4-Ethylaniline
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 216
Melting Pt: –5 °C
Density: 0.96875 g/cm³ (20 °C)
Flash Pt: 97
Storage Temperature: Ambient
MDL Number: MFCD00007921
CAS Number: 589-16-2
EINECS: 209-637-0
UN: 2810
ADR: 6.1 III