4-Hexylaniline, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
233501-25G 233501-5G
SIAL233501-25GEA 285 EUR
SIAL233501-25G SIAL233501-5G
4-Hexylaniline, Sigma-Aldrich®
4-Hexylaniline
Formula: C₁₂H₁₉N
MW: 177.29 g/mol
Boiling Pt: 96 °C (0.6 mmHg)
Density: 0.91 g/cm³ (20 °C)
MDL Number: MFCD00007927
CAS Number: 33228-45-4
EINECS: 251-409-8