4-Methoxybenzylamine, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 4-(Aminomethyl)anisole, p-Anisylamine

M-Clarity™ quality level = MQ200

M11103-100G M11103-25G
SIALM11103-100GEA 481 EUR
SIALM11103-100G SIALM11103-25G
4-Methoxybenzylamine, Sigma-Aldrich®
4-Methoxybenzylamine
Formula: C₈H₁₁NO
MW: 137.18 g/mol
Boiling Pt: 236 °C (1013 hPa)
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008122
CAS Number: 2393-23-9
EINECS: 219-247-2
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR