2,6-Diaminotoluene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1,3-Diamino-2-methylbenzene, Toluene-2,6-diamine
M-Clarity™ quality level = MQ100
148113-5G 148113-25G
SIAL148113-5GEA 21 EUR
SIAL148113-5G SIAL148113-25G
2,6-Diaminotoluene, Sigma-Aldrich®
2,6-Diaminotoluene
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 280…282 °C (1013 hPa)
Melting Pt: 113 °C
MDL Number: MFCD00007800
CAS Number: 823-40-5
EINECS: 212-513-9
UN: 2811
ADR: 6.1,III