2-Nitrobenzyl alcohol 97%
Supplier: Thermo Scientific
Warning
Synonyms: (2-Nitrophenyl)methanol
128510250 128511000 128510050
ACRO128510250EA 52.3 EUR
ACRO128510250 ACRO128511000 ACRO128510050
2-Nitrobenzyl alcohol 97%
2-Nitrobenzyl alcohol
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 270 °C (760 mmHg)
Melting Pt: 69…73 °C
Storage Temperature: Ambient
MDL Number: MFCD00007186
CAS Number: 612-25-9
EINECS: 210-302-6
UN: 2811
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Beige to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 69 to 73 °C
GC ≥96.0 %