4-Nitrobenzyl alcohol

Supplier: Thermo Scientific

Synonyms: (4-Nitrophenyl)methanol

SB01165.1 SB01165.10 SB01165.50
MAYBSB01165.1EA 0 EUR
MAYBSB01165.1 MAYBSB01165.10 MAYBSB01165.50
4-Nitrobenzyl alcohol
4-Nitrobenzyl alcohol
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 321 °C (1013 hPa)
Melting Pt: 90…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007376
CAS Number: 619-73-8
EINECS: 210-611-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR