4-Nitrobenzyl alcohol, Sigma-Aldrich®

Supplier: Merck

Synonyms: (4-Nitrophenyl)methanol

M-Clarity™ quality level = MQ100

N12821-100G N12821-25G
SIALN12821-100GEA 117 EUR
SIALN12821-100G SIALN12821-25G
4-Nitrobenzyl alcohol, Sigma-Aldrich®
4-Nitrobenzyl alcohol
Formula: C₇H₇NO₃
MW: 153.14 g/mol
Boiling Pt: 321 °C (1013 hPa)
Melting Pt: 90…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00007376
CAS Number: 619-73-8
EINECS: 210-611-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR