4-Methylanisole, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
148091-100ML 148091-1L
SIAL148091-100MLEA 25.2 EUR
SIAL148091-100ML SIAL148091-1L
4-Methylanisole, Sigma-Aldrich®
4-Methylanisole
Formula: C₈H₁₀O
Boiling Pt: 174 °C (1013 hPa)
Melting Pt: –36 °C
Density: 0.97 g/cm³ (20 °C)
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00008413
CAS Number: 104-93-8
EINECS: 203-253-7
UN: 3271
ADR: 3,III