2-Methoxy-p-cresol, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Creosol, 1-Hydroxy-2-methoxy-4-methylbenzene, 2-Methoxy-p-cresol, 2-Methoxy-4-methylphenol, 4-Hydroxy-3-methoxytoluene, 4-Methylguaiacol, 2-Hydroxy-5-methylanisol
M-Clarity™ quality level = MQ300
W267104-100G-K
W267104-10KG-K
W267104-1KG-K
SIALW267104-100G-KEA
76.2
EUR
SIALW267104-100G-K
SIALW267104-10KG-K
SIALW267104-1KG-K
2-Methoxy-p-cresol, Sigma-Aldrich®
2-Methoxy-p-cresol
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol Boiling Pt: 219…222 °C (1013 hPa) Density: 1.09 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002378 CAS Number: 93-51-6 EINECS: 202-252-9 Merck Index: 13,02599 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...