2-Methoxy-p-cresol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Creosol, 1-Hydroxy-2-methoxy-4-methylbenzene, 2-Methoxy-p-cresol, 2-Methoxy-4-methylphenol, 4-Hydroxy-3-methoxytoluene, 4-Methylguaiacol, 2-Hydroxy-5-methylanisol
M-Clarity™ quality level = MQ300
W267112-100G-K W267112-1KG-K
SIALW267112-100G-KEA 308 EUR
SIALW267112-100G-K SIALW267112-1KG-K
2-Methoxy-p-cresol, Sigma-Aldrich®
2-Methoxy-p-cresol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 219…222 °C (1013 hPa)
Density: 1.09 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002378
CAS Number: 93-51-6
EINECS: 202-252-9
Merck Index: 13,02599