4-Hydroxy-3-methoxybenzyl alcohol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Vanillyl alcohol, 4-(Hydroxymethyl)-2-methoxyphenol, 2-Hydroxy-5-(hydroxymethyl)anisole, (4-Hydroxy-3-methoxyphenyl)methanol
M-Clarity™ quality level = MQ100
175536-50G 175536-250G
SIAL175536-50GEA 61.1 EUR
SIAL175536-50G SIAL175536-250G
4-Hydroxy-3-methoxybenzyl alcohol, Sigma-Aldrich®
4-Hydroxy-3-methoxybenzyl alcohol
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 293
Melting Pt: 113
MDL Number: MFCD00004659
CAS Number: 498-00-0
EINECS: 207-852-4