4-Hydroxy-3-methoxybenzyl alcohol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Vanillyl alcohol, 4-(Hydroxymethyl)-2-methoxyphenol, 2-Hydroxy-5-(hydroxymethyl)anisole, (4-Hydroxy-3-methoxyphenyl)methanol
M-Clarity™ quality level = MQ300
W373702-100G-K W373702-1KG-K
SIALW373702-100G-KEA 111 EUR
SIALW373702-100G-K SIALW373702-1KG-K
4-Hydroxy-3-methoxybenzyl alcohol, Sigma-Aldrich®
4-Hydroxy-3-methoxybenzyl alcohol
Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 293
Melting Pt: 113
MDL Number: MFCD00004659
CAS Number: 498-00-0
EINECS: 207-852-4