2-Methyl-1-phenyl-1-propanol ≥98%
Supplier: Thermo Scientific
Warning
Synonyms: α-Isopropylbenzyl alcohol, 2-Methyl-1-phenylpropan-1-ol
B20729.06 B20729.14
B20729.06EA 66.9 EUR
B20729.06 B20729.14
2-Methyl-1-phenyl-1-propanol ≥98%
2-Methyl-1-phenyl-1-propanol
Purity: 98% CAS No.: 611-69-8 Molecular Formula: C10H14O Formula Weight: 150.22 Flash Point: 86°(187°F) Refractive Index: 1.5130 Boiling Point: 124-125°/15mm MDL No.: MFCD00065000
Formula: C₆H₅CH(OH)CH(CH₃)₂
MW: 150.22 g/mol
Boiling Pt: 124…125 °C (15 mmHg)
Melting Pt: –14 °C
Density: 0.964
Flash Pt: 86 °C (187 °F)
Storage Temperature: Ambient
MDL Number: MFCD00065000
CAS Number: 611-69-8
EINECS: 210-274-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=954485764810b5712627a8e76df6a8dd392a6e2a&vid=954485764810b5712627a8e76df6a8dd392a6e2a&prid=web&clid=SAPDAM