3-Phenyl-1-propanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-Phenyl-1-Propanol
M-Clarity™ quality level = MQ100
140856-250G 140856-50G
SIAL140856-250GEA 54.7 EUR
SIAL140856-250G SIAL140856-50G
3-Phenyl-1-propanol, Sigma-Aldrich®
3-Phenyl-1-propanol
Formula: (C₆H₅)CH₂CH₂CH₂OH
MW: 136.19 g/mol
Boiling Pt: 455 °C (1013 hPa)
Melting Pt: –18 °C
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002950
CAS Number: 122-97-4
EINECS: 204-587-6