3-Phenyl-1-propanol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
3-Phenyl-1-Propanol
M-Clarity™ quality level = MQ300
W288500-10KG
W288500-1KG
W288500-5KG
W288500-SAMPLE
SIALW288500-10KGEA
807
EUR
SIALW288500-10KG
SIALW288500-1KG
SIALW288500-5KG
SIALW288500-SAMPLE
3-Phenyl-1-propanol, Sigma-Aldrich®
3-Phenyl-1-propanol
Formula:
(C₆H₅)CH₂CH₂CH₂OH MW: 136.19 g/mol Boiling Pt: 455 °C (1013 hPa) Melting Pt: –18 °C Density: 1.01 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002950 CAS Number: 122-97-4 EINECS: 204-587-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...