2-tert-Butylphenol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
B99405-500ML B99405-50ML
SIALB99405-500MLEA 109 EUR
SIALB99405-500ML SIALB99405-50ML
2-tert-Butylphenol, Sigma-Aldrich®
2-tert-Butylphenol
Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 221 °C (1013 hPa)
Melting Pt: –7 °C
Density: 0.9823 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002223
CAS Number: 88-18-6
EINECS: 201-807-2
UN: 3145
ADR: 8,III