2-Ethylphenol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
SIALE44000-100GEA
172
EUR
SIALE44000-100G
SIALE44000-25G
SIALE44000-5G
2-Ethylphenol, Sigma-Aldrich®
2-Ethylphenol
Formula:
C₈H₁₀O MW: 122,17 g/mol Boiling Pt: 204,5 °C (1013 hPa) Melting Pt: –18 °C Density: 1,0371 g/cm³ (0 °C) |
MDL Number:
MFCD00002249 UN: 3145 ADR: 8,III Merck Index: 13,03871 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...