2-Isopropylphenol, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
129526-100G 129526-5G
SIAL129526-100GEA 61.3 EUR
SIAL129526-100G SIAL129526-5G
2-Isopropylphenol, Sigma-Aldrich®
2-Isopropylphenol
Formula: C₉H₁₂O
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: 14…16 °C
Density: 0.995 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002224
CAS Number: 88-69-7
EINECS: 201-852-8
UN: 3145
ADR: 8,II