2-Isopropylphenol, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ300
W346101-10KG W346101-1KG W346101-25KG W346101-SAMPLE
SIALW346101-10KGEA 1600 EUR
SIALW346101-10KG SIALW346101-1KG SIALW346101-25KG SIALW346101-SAMPLE
2-Isopropylphenol, Sigma-Aldrich®
2-Isopropylphenol
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 213
Melting Pt: 14…16 °C
Density: 0.995 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002224
CAS Number: 88-69-7
EINECS: 201-852-8
UN: 3145
ADR: 8 II