o-Cresol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1-Hydroxy-2-methylbenzene, 2-Methylphenol, 2-Hydroxytoluene
M-Clarity™ quality level = MQ300
W348007-10KG-K W348007-1KG-K W348007-25KG-K
SIALW348007-10KG-KEA 379 EUR
SIALW348007-10KG-K SIALW348007-1KG-K SIALW348007-25KG-K
o-Cresol, Sigma-Aldrich®
o-Cresol
Formula: C₇H₈O
MW: 108.14 g/mol
Boiling Pt: 191 °C (1013 hPa)
Melting Pt: 30…34 °C
Density: 1.047 g/cm³ (20 °C)
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00002226
CAS Number: 95-48-7
EINECS: 202-423-8
UN: 3455
ADR: 6.1,II
Merck Index: 13,02604