4-Isopropylphenol, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ100

175404-100G 175404-5G
SIAL175404-100GEA 99.5 EUR
SIAL175404-100G SIAL175404-5G
4-Isopropylphenol, Sigma-Aldrich®
4-Isopropylphenol
Formula: C₉H₁₂O
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: 60 °C
Density: 0.99 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002372
CAS Number: 99-89-8
EINECS: 202-798-8
UN: 2430
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR