2-Methylresorcinol, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2,6-Dihydroxytoluene, 2-Methylbenzene-1,3-diol

M-Clarity™ quality level = MQ100

302589-100G 302589-25G
SIAL302589-100GEA 151 EUR
SIAL302589-100G SIAL302589-25G
2-Methylresorcinol, Sigma-Aldrich®
2-Methylresorcinol
Formula: C₇H₈O₂
Boiling Pt: 264 °C (1013 hPa)
Melting Pt: 120…122 °C
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00002271
CAS Number: 608-25-3
EINECS: 210-155-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR