4-Hexylresorcinol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Hexylresorcinol, 4-Hexylbenzene-1,3-diol
M-Clarity™ quality level = MQ100
209465-100G 209465-25G
SIAL209465-100GEA 205 EUR
SIAL209465-100G SIAL209465-25G
4-Hexylresorcinol, Sigma-Aldrich®
4-Hexylresorcinol
Formula: C₁₂H₁₈O₂
MW: 194.27 g/mol
Boiling Pt: 333
Melting Pt: 64…67 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00002284
CAS Number: 136-77-6
EINECS: 205-257-4
Merck Index: 12,04750