4-Hydroxybenzyl alcohol, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
H20806-100G H20806-10G H20806-25G
SIALH20806-100GEA 149 EUR
SIALH20806-100G SIALH20806-10G SIALH20806-25G
4-Hydroxybenzyl alcohol, Sigma-Aldrich®
4-Hydroxybenzyl alcohol
Formula: C₇H₈O₂
Boiling Pt: 252 °C (1013 hPa)
Melting Pt: 115…118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004658
CAS Number: 623-05-2
EINECS: 210-768-0