4-Hydroxybenzyl alcohol, Sigma-Aldrich®

Supplier: Merck

Synonyms: (4-Hydroxyphenyl)methanol, p-Hydroxybenzyl Alcohol, 4-(Hydroxymethyl)phenol

M-Clarity™ quality level = MQ300

W398705-100G W398705-1KG W398705-5KG W398705-SAMPLE
SIALW398705-100GEA 135 EUR
SIALW398705-100G SIALW398705-1KG SIALW398705-5KG SIALW398705-SAMPLE
4-Hydroxybenzyl alcohol, Sigma-Aldrich®
4-Hydroxybenzyl alcohol
Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 252
Melting Pt: 115…118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004658
CAS Number: 623-05-2
EINECS: 210-768-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR