4-tert-Butylthiophenol
Supplier: Thermo Scientific
Warning
TL00626.1 TL00626.10
MAYBTL00626.1EA 0 EUR
MAYBTL00626.1 MAYBTL00626.10
4-tert-Butylthiophenol
4-tert-Butylthiophenol
Formula: C₁₀H₁₄S
Boiling Pt: 238 °C (1013 hPa)
Melting Pt: –11 °C
Density: 0.9917 g/cm³ (16 °C)
MDL Number: MFCD00022067
CAS Number: 2396-68-1
EINECS: 219-255-6
UN: 3334
ADR: 9,