4-tert-Butylthiophenol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-tert-Butyl-4-mercaptobenzene, 4-tert-Butylbenzene-1-thiol, 4-tert-Butylbenzenethiol
M-Clarity™ quality level = MQ200
593656-25G 593656-5G
SIAL593656-25GEA 264 EUR
SIAL593656-25G SIAL593656-5G
4-tert-Butylthiophenol, Sigma-Aldrich®
4-tert-Butylthiophenol
Formula: C₁₀H₁₄S
MW: 166.29 g/mol
Boiling Pt: 238
Melting Pt: –11 °C
Density: 0.9917 g/cm³ (16 °C)
MDL Number: MFCD00022067
CAS Number: 2396-68-1
EINECS: 219-255-6
UN: 3334
ADR: 9