2-Toluenethiol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Thiocresol, 1-Mercapto-2-methylbenzene, o-Toluenethiol, 2-Methylbenzene-1-thiol, 2-Methylbenzenethiol, o-Thiocresol, 2-Methylthiophenol
MQ300
W324000-100G-K W324000-1KG-K W324000-500G-K
SIALW324000-100G-KEA 452 EUR
SIALW324000-100G-K SIALW324000-1KG-K SIALW324000-500G-K
2-Toluenethiol, Sigma-Aldrich®
2-Toluenethiol
Formula: C₇H₈S
MW: 124.21 g/mol
Boiling Pt: 195
Melting Pt: 4.5
Density: 1.041 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004838
CAS Number: 137-06-4
EINECS: 205-276-8
UN: 2810
ADR: 6.1 III
Merck Index: 12,09461