Toluene-2-sulphonamide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Methylbenzene-1-sulphonamide, 2-Methylbenzenesulphonamide, o-Toluenesulphonamide
M-Clarity™ quality level = MQ200
257990-100G 257990-25G
SIAL257990-100GEA 194 EUR
SIAL257990-100G SIAL257990-25G
Toluene-2-sulphonamide, Sigma-Aldrich®
Toluene-2-sulphonamide
Formula: C₇H₉NO₂S
MW: 171.22 g/mol
Boiling Pt: 330 °C (1013 hPa)
Melting Pt: 155…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00007934
CAS Number: 88-19-7
EINECS: 201-808-8