p-Toluenesulphonyl hydrazide, Sigma-Aldrich®
Supplier: Merck
Synonyms: 4-Methylbenzenesulphonylhydrazide, 4-Toluenesulphonyl hydrazide, 4-Methylbenzenesulphonhydrazide, 4-Methylbenzenesulphonic acid hydrazide, 4-Methylbenzenesulphonohydrazide, Toluene-4-sulphonohydrazide
132004-100G 132004-25G 132004-500G
SIAL132004-100GEA 91.5 EUR
SIAL132004-100G SIAL132004-25G SIAL132004-500G
p-Toluenesulphonyl hydrazide, Sigma-Aldrich®
p-Toluenesulphonyl hydrazide
Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 105…108 °C
Density: 1,4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007588
CAS Number: 1576-35-8
EINECS: 216-407-3