Ethyl toluene-4-sulphonate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Ethyl 4-toluenesulphonate, Ethyl p-toluenesulphonate, p-Toluenesulphonic acid ethyl ester, Ethyl 4-methylbenzenesulphonate, Toluene-4-sulphonic acid ethyl ester
M-Clarity™ quality level = MQ200
104256-1KG 104256-250G 104256-50G
SIAL104256-1KGEA 209 EUR
SIAL104256-1KG SIAL104256-250G SIAL104256-50G
Ethyl toluene-4-sulphonate, Sigma-Aldrich®
Ethyl toluene-4-sulphonate
Formula: C₉H₁₂O₃S
MW: 200.26 g/mol
Boiling Pt: 158
Melting Pt: 30…32 °C
Density: 1.174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009100
CAS Number: 80-40-0
EINECS: 201-276-7
UN: 2811
ADR: 6.1 III
Merck Index: 13,03889