Methyl toluene-4-sulphonate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Methyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid methyl ester, Methyl p-toluenesulphonate, Toluene-4-sulphonic acid methyl ester
M-Clarity™ quality level = MQ200
158992-100G 158992-500G
SIAL158992-100GEA 38.1 EUR
SIAL158992-100G SIAL158992-500G
Methyl toluene-4-sulphonate, Sigma-Aldrich®
Methyl toluene-4-sulphonate
Formula: C₈H₁₀O₃S
MW: 186.23 g/mol
Boiling Pt: 292
Melting Pt: 28…30 °C
Density: 1.23 g/cm³ (20 °C)
Flash Pt: 130
Storage Temperature: Ambient
MDL Number: MFCD00008417
CAS Number: 80-48-8
EINECS: 201-283-5