sec-Butylbenzene, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Phenylbutane

M-Clarity™ quality level = MQ100

B90408-100ML B90408-500ML
SIALB90408-100MLEA 72.4 EUR
SIALB90408-100ML SIALB90408-500ML
sec-Butylbenzene, Sigma-Aldrich®
sec-Butylbenzene
Formula: C₆H₅CH(CH₃)CH₂CH₃
MW: 134.22 g/mol
Boiling Pt: 173 °C (1013 hPa)
Melting Pt: –75 °C
Density: 0.861 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009329
CAS Number: 135-98-8
EINECS: 205-227-0
UN: 2709
ADR: 3,III
Merck Index: 13,01549
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bb1eb7754ba9f4f037280b9010ef49cadd1a43c7&vid=d32e383af74575be997125b4f7121f54764ab833&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR