n-Hexylbenzene, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1-Phenylhexane, Hexylbenzene

M-Clarity™ quality level = MQ200

P25701-100G P25701-25G
SIALP25701-100GEA 190 EUR
SIALP25701-100G SIALP25701-25G
n-Hexylbenzene, Sigma-Aldrich®
n-Hexylbenzene
Formula: C₁₂H₁₈
MW: 162.27 g/mol
Boiling Pt: 226
Melting Pt: –61 °C
Density: 0.8666 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009526
CAS Number: 1077-16-3
EINECS: 214-070-7
UN: 3082
ADR: 9 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR