Bromocresol purple, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
5',5''-Dibromo-o-cresolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
17490-10G
17490-50G
SIAL17490-10GEA
59.8
EUR
SIAL17490-10G
SIAL17490-50G
Bromocresol purple, Sigma-Aldrich®
Bromocresol purple
Formula:
C₂₁H₁₆Br₂O₅S MW: 540.23 g/mol Melting Pt: 242 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011681 CAS Number: 115-40-2 EINECS: 204-087-8 Merck Index: 13,01371 |